Assay Details
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052877.4 | NP_443109.2 | 7 - 8 | 857 | 71 | |
GenBank mRNA | BC010543.1 | - | 6 - 7 | 798 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002584.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01040196.hg.1 | TC01002584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01040196.hg.1 | ENST00000290663.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040196.hg.1 | ENST00000372457.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040196.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01040195.hg.1 | TC01002584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01040195.hg.1 | ENST00000290663.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040195.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204150.hg.1 | TC0100013925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204148.hg.1 | TC0100013925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204149.hg.1 | TC0100013925.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 175513 | - | - |
