Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041200.hg.1 |
TC01002634.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041200.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041198.hg.1 |
TC01002634.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041198.hg.1 |
ENST00000371865.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041198.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041201.hg.1 |
TC01002634.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041201.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000438589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000438589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205596.hg.1 |
TC0100014109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000438589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000438589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014109.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
ENST00000435576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205593.hg.1 |
TC0100014109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205600.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205600.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205600.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205600.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205600.hg.1 |
TC0100014109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205597.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205597.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205597.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205597.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205597.hg.1 |
TC0100014109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205598.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205598.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205598.hg.1 |
ENST00000606738.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205598.hg.1 |
NM_001194986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205598.hg.1 |
TC0100014109.hg.1 |
Clariom D TC |