Assay Details
Gene Symbol: FAM96A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032231.5 | NP_115607.1 | 2 - 3 | 530 | 118 | |
NR_110310.1 | 3 - 4 | 648 | 118 | |||
GenBank mRNA | AK312171.1 | - | 2 - 3 | 472 | 118 | |
BC008865.2 | - | 2 - 3 | 432 | 4128736 | 118 | |
CN421575.1 | - | 2 - 3 | 458 | 118 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001547.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15014664.hg.1 | ENST00000300030.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15014664.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15014664.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15014664.hg.1 | TC15001547.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500171881.hg.1 | TC1500009731.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500171883.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171883.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500171883.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500171883.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500171883.hg.1 | TC1500009731.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000300030.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000380290.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000557835.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000558779.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | ENST00000559950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001014812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_001289108.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NM_032231.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009731.hg.1 | NR_110310.1 | Refseq |
