Assay Details
| Gene Symbol: FGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004356.2 | NP_001004356.1 | 4 - 5 | 577 | 111 | |
| NM_001004358.1 | NP_001004358.1 | 4 - 5 | 460 | 111 | ||
| NM_021923.3 | NP_068742.2 | 3 - 4 | 450 | 111 | ||
| XM_011513486.1 | XP_011511788.1 | 4 - 5 | 581 | 111 | ||
| GenBank mRNA | AF279689.1 | - | 4 - 5 | 460 | 111 | |
| AF312678.1 | - | 577 | 111 | |||
| AJ277437.1 | - | 3 - 4 | 450 | 111 | ||
| AK172829.1 | - | 3 - 4 | 434 | 111 | ||
| AK225486.1 | - | 3 - 4 | 421 | 111 | ||
| AK314365.1 | - | 4 - 5 | 578 | 111 | ||
| AY358303.1 | - | 577 | 111 | |||
| BC036769.1 | - | 4 - 5 | 589 | 5744684 | 111 | |
| BQ689625.1 | - | 3 - 4 | 405 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000016.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
heart valve morphogenesis
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
substrate adhesion-dependent cell spreading
protein homooligomerization
protein heterooligomerization
ventricular septum morphogenesis
diaphragm development
cell-cell adhesion via plasma-membrane adhesion molecules
fibroblast growth factor-activated receptor activity
heparin binding
fibroblast growth factor binding
skeletal system development
heart valve morphogenesis
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
substrate adhesion-dependent cell spreading
protein homooligomerization
protein heterooligomerization
ventricular septum morphogenesis
diaphragm development
cell-cell adhesion via plasma-membrane adhesion molecules
fibroblast growth factor-activated receptor activity
heparin binding
fibroblast growth factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | TC04000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | ENST00000398484.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | ENST00000504138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | ENST00000510644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | ENST00000542622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04000426.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400144598.hg.1 | TC0400006493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |

FGFRL1
Genomic Map