Assay Details
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004356.2 | NP_001004356.1 | 5 - 6 | 868 | 62 | |
NM_001004358.1 | NP_001004358.1 | 5 - 6 | 751 | 62 | ||
NM_021923.3 | NP_068742.2 | 4 - 5 | 741 | 62 | ||
XM_011513486.1 | XP_011511788.1 | 5 - 6 | 872 | 62 | ||
GenBank mRNA | AF279689.1 | - | 5 - 6 | 751 | 62 | |
AF312678.1 | - | 868 | 62 | |||
AJ277437.1 | - | 4 - 5 | 741 | 62 | ||
AK172829.1 | - | 4 - 5 | 725 | 62 | ||
AK225486.1 | - | 4 - 5 | 712 | 62 | ||
AK314365.1 | - | 5 - 6 | 869 | 62 | ||
AY358303.1 | - | 868 | 62 | |||
BC015228.1 | - | 1 - 2 | 152 | 62 | ||
BC036769.1 | - | 5 - 6 | 880 | 5744684 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000016.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
heart valve morphogenesis
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
substrate adhesion-dependent cell spreading
protein homooligomerization
protein heterooligomerization
ventricular septum morphogenesis
diaphragm development
cell-cell adhesion via plasma-membrane adhesion molecules
fibroblast growth factor-activated receptor activity
heparin binding
fibroblast growth factor binding
skeletal system development
heart valve morphogenesis
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
substrate adhesion-dependent cell spreading
protein homooligomerization
protein heterooligomerization
ventricular septum morphogenesis
diaphragm development
cell-cell adhesion via plasma-membrane adhesion molecules
fibroblast growth factor-activated receptor activity
heparin binding
fibroblast growth factor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | TC04000016.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | ENST00000398484.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | ENST00000504138.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | ENST00000510644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | ENST00000542622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04000427.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400144599.hg.1 | TC0400006493.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000264748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000398484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000504138.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000507339.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000510644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | ENST00000512562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004356.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_001004358.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006493.hg.1 | NM_021923.3 | Refseq |
