Assay Details
| Gene Symbol: FIZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527426.2 | XP_011525728.1 | 2 - 3 | 344 | 73 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000221665.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000587678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000590714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | ENST00000592585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011484.hg.1 | NM_032836.2 | Refseq |

FIZ1
Genomic Map