Assay Details
Gene Symbol: MMRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007351.2 | NP_031377.2 | 1 - 2 | 696 | 92 | |
XM_005262856.2 | XP_005262913.1 | 1 - 2 | 721 | 92 | ||
XM_017007893.1 | XP_016863382.1 | 2 - 3 | 937 | 92 | ||
GenBank mRNA | AK313566.1 | - | 1 - 2 | 721 | 92 | |
BC063848.1 | - | 1 - 2 | 696 | 6180674 | 92 | |
U27109.1 | - | 1 - 2 | 696 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000493.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04007162.hg.1 | TC04000493.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04007162.hg.1 | ENST00000264790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007162.hg.1 | ENST00000394980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007162.hg.1 | ENST00000394981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007162.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007162.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04007156.hg.1 | TC04000493.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04007156.hg.1 | ENST00000264790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007156.hg.1 | ENST00000394980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007156.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04007158.hg.1 | TC04000493.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04007158.hg.1 | ENST00000264790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007158.hg.1 | ENST00000394980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007158.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04007159.hg.1 | TC04000493.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04007159.hg.1 | ENST00000264790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007159.hg.1 | ENST00000394980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007159.hg.1 | ENST00000394981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007159.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04007157.hg.1 | TC04000493.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04007157.hg.1 | ENST00000264790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007157.hg.1 | ENST00000394980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007157.hg.1 | ENST00000394981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04007157.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156012.hg.1 | TC0400008134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156013.hg.1 | TC0400008134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400156015.hg.1 | TC0400008134.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000264790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000394980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000506328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | ENST00000508372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008134.hg.1 | NM_007351.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 128509 | - | - |
