Assay Details
Gene Symbol: SPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012444.2 | NP_036576.1 | 2 - 3 | 329 | 89 | |
XM_005260379.3 | XP_005260436.1 | 2 - 3 | 347 | 89 | ||
XM_005260380.3 | XP_005260437.1 | 2 - 3 | 347 | 89 | ||
XM_005260381.3 | XP_005260438.1 | 2 - 3 | 347 | 89 | ||
XM_005260382.4 | XP_005260439.1 | 2 - 3 | 347 | 89 | ||
XM_011528757.2 | XP_011527059.1 | 2 - 3 | 181 | 89 | ||
GenBank mRNA | AF149310.1 | - | 2 - 3 | 269 | 89 | |
AF169385.1 | - | 2 - 3 | 344 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528757.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
