Assay Details
| Gene Symbol: TRMT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202423.1 | NP_001189352.1 | 2 - 3 | 225 | 106 | |
| NM_030934.4 | NP_112196.3 | 2 - 3 | 617 | 106 | ||
| XM_011510028.2 | XP_011508330.1 | 2 - 3 | 2054 | 106 | ||
| XM_017002419.1 | XP_016857908.1 | 2 - 3 | 589 | 106 | ||
| GenBank mRNA | AF288399.1 | - | 2 - 3 | 539 | 106 | |
| AK074936.1 | - | 2 - 3 | 566 | 106 | ||
| AK074993.1 | - | 2 - 3 | 589 | 106 | ||
| AK098359.1 | - | 2 - 3 | 584 | 106 | ||
| AK131460.1 | - | 1 - 2 | 1259 | 106 | ||
| AK226103.1 | - | 2 - 3 | 332 | 106 | ||
| AK302166.1 | - | 2 - 3 | 149 | 106 | ||
| AK312801.1 | - | 2 - 3 | 586 | 106 | ||
| BC035430.1 | - | 2 - 3 | 569 | 106 | ||
| BC039738.1 | - | 2 - 3 | 580 | 106 | ||
| BC045535.1 | - | 2 - 3 | 566 | 5296208 | 106 | |
| BC053961.1 | - | 2 - 3 | 566 | 106 | ||
| DC398320.1 | - | 2 - 3 | 225 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003627.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK131460.1,AK302166.1,NM_001202423.1,XM_011510028.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055796.hg.1 | TC01003627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055796.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000465827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000465827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000465827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055792.hg.1 | TC01003627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055792.hg.1 | ENST00000367504.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055792.hg.1 | ENST00000367506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055792.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055793.hg.1 | TC01003627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055793.hg.1 | ENST00000367504.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055793.hg.1 | ENST00000367506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055793.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055795.hg.1 | TC01003627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055795.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055790.hg.1 | TC01003627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055790.hg.1 | ENST00000367504.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055790.hg.1 | ENST00000367506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055790.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055790.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055791.hg.1 | TC01003627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055791.hg.1 | ENST00000367504.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055791.hg.1 | ENST00000367506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055791.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055791.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226575.hg.1 | TC0100016685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226576.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226576.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226576.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226576.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226576.hg.1 | TC0100016685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226581.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226581.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226581.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226581.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226581.hg.1 | TC0100016685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226574.hg.1 | TC0100016685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226577.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226577.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226577.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226577.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226577.hg.1 | TC0100016685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226582.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226582.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226582.hg.1 | TC0100016685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000458395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000465827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000465827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000465827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | ENST00000487028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_001202423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016685.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226578.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226578.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226578.hg.1 | ENST00000367506.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226578.hg.1 | NM_030934.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226578.hg.1 | TC0100016685.hg.1 | Clariom D TC |

TRMT1L
Genomic Map