Assay Details
Gene Symbol: CDC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024529.4 | NP_078805.3 | 1 - 2 | 375 | 62 | |
XM_006711537.3 | XP_006711600.1 | 1 - 2 | 299 | 62 | ||
GenBank mRNA | AF312865.1 | - | 1 - 2 | 140 | 62 | |
AK026969.1 | - | 1 - 2 | 267 | 62 | ||
AK226038.1 | - | 1 - 2 | 272 | 62 | ||
AK300929.1 | - | 1 - 2 | 234 | 62 | ||
AK314772.1 | - | 1 - 2 | 133 | 62 | ||
BC007325.2 | - | 1 - 2 | 158 | 62 | ||
BC065037.1 | - | 1 - 2 | 212 | 6170851 | 62 | |
BM471846.1 | - | 1 - 2 | 328 | 62 | ||
DC298135.1 | - | 1 - 2 | 375 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001626.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK026969.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
transcription elongation from RNA polymerase II promoter
mRNA polyadenylation
cell cycle
negative regulation of cell proliferation
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of Wnt signaling pathway
positive regulation of mRNA 3'-end processing
protein destabilization
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
cellular response to lipopolysaccharide
beta-catenin-TCF complex assembly
negative regulation of G1/S transition of mitotic cell cycle
RNA polymerase II core binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate commitment
transcription elongation from RNA polymerase II promoter
mRNA polyadenylation
cell cycle
negative regulation of cell proliferation
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of Wnt signaling pathway
positive regulation of mRNA 3'-end processing
protein destabilization
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
cellular response to lipopolysaccharide
beta-catenin-TCF complex assembly
negative regulation of G1/S transition of mitotic cell cycle
RNA polymerase II core binding
protein binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01025131.hg.1 | TC01001626.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01025131.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025131.hg.1 | ENST00000445394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025131.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01025129.hg.1 | TC01001626.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01025129.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025129.hg.1 | ENST00000445394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025129.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01025127.hg.1 | TC01001626.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01025127.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025127.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01025126.hg.1 | TC01001626.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01025126.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180929.hg.1 | TC0100011027.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000477868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | ENST00000482484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011027.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180925.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180925.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180925.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180925.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180925.hg.1 | TC0100011027.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100180924.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180924.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180924.hg.1 | TC0100011027.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100180927.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180927.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180927.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180927.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180927.hg.1 | TC0100011027.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100180926.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180926.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180926.hg.1 | ENST00000367435.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100180926.hg.1 | NM_024529.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100180926.hg.1 | TC0100011027.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 205190 | - | - |
