Assay Details
| Gene Symbol: SHROOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001649.3 | NP_001640.1 | 1 - 2 | 258 | 51 | |
| XM_005274500.4 | XP_005274557.2 | 1 - 2 | 308 | 51 | ||
| GenBank mRNA | BC140866.1 | - | 1 - 2 | 219 | 9021734 | 51 |
| X83543.1 | - | 1 - 2 | 258 | 51 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000034.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
lens morphogenesis in camera-type eye
brain development
eye pigment granule organization
cell migration
establishment of melanosome localization
melanosome organization
sodium ion transmembrane transport
camera-type eye development
cellular pigment accumulation
ear development
apical protein localization
camera-type eye morphogenesis
actin filament bundle assembly
actin binding
protein binding
beta-catenin binding
ligand-gated sodium channel activity
actin filament binding
cell morphogenesis
lens morphogenesis in camera-type eye
brain development
eye pigment granule organization
cell migration
establishment of melanosome localization
melanosome organization
sodium ion transmembrane transport
camera-type eye development
cellular pigment accumulation
ear development
apical protein localization
camera-type eye morphogenesis
actin filament bundle assembly
actin binding
protein binding
beta-catenin binding
ligand-gated sodium channel activity
actin filament binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000447.hg.1 | TC0X000034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000447.hg.1 | ENST00000380913.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000447.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000493668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000493668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | ENST00000493668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000448.hg.1 | TC0X000034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000448.hg.1 | ENST00000380913.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000448.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000446.hg.1 | TC0X000034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000446.hg.1 | ENST00000380913.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000446.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144985.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144985.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144985.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144985.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144985.hg.1 | TC0X00006585.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000418909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000452575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000493668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000493668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | ENST00000493668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006585.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144982.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144982.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144982.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144982.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144982.hg.1 | TC0X00006585.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00144983.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144983.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144983.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144983.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144983.hg.1 | TC0X00006585.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00144984.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144984.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144984.hg.1 | ENST00000380913.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144984.hg.1 | NM_001649.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144984.hg.1 | TC0X00006585.hg.1 | Clariom D TC |

SHROOM2
Genomic Map