| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000449.hg.1 |
TC0X000034.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000449.hg.1 |
ENST00000380913.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X000449.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000493668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000493668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
ENST00000493668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000418909.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000452575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000493668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000493668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
ENST00000493668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006585.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144987.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144987.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144987.hg.1 |
ENST00000380913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00144987.hg.1 |
NM_001649.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00144987.hg.1 |
TC0X00006585.hg.1 |
Clariom D TC |