Assay Details
| Gene Symbol: GNPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014236.3 | NP_055051.1 | 2 - 3 | 469 | 148 | |
| XM_005273313.4 | XP_005273370.1 | 2 - 3 | 429 | 148 | ||
| GenBank mRNA | AB209882.1 | - | 2 - 3 | 374 | 148 | |
| AF043937.1 | - | 2 - 3 | 345 | 148 | ||
| AJ002190.1 | - | 2 - 3 | 420 | 148 | ||
| AK222773.1 | - | 2 - 3 | 425 | 148 | ||
| AK223304.1 | - | 2 - 3 | 428 | 148 | ||
| AK314181.1 | - | 2 - 3 | 432 | 148 | ||
| BC000450.2 | - | 2 - 3 | 316 | 2821393 | 148 | |
| DA552987.1 | - | 2 - 3 | 471 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001915.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycerophospholipid metabolic process
phosphatidic acid biosynthetic process
synapse assembly
response to nutrient
ether lipid biosynthetic process
cerebellum morphogenesis
paranodal junction assembly
response to drug
response to starvation
membrane organization
response to fatty acid
receptor binding
glycerone-phosphate O-acyltransferase activity
palmitoyl-CoA hydrolase activity
glycerophospholipid metabolic process
phosphatidic acid biosynthetic process
synapse assembly
response to nutrient
ether lipid biosynthetic process
cerebellum morphogenesis
paranodal junction assembly
response to drug
response to starvation
membrane organization
response to fatty acid
receptor binding
glycerone-phosphate O-acyltransferase activity
palmitoyl-CoA hydrolase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010522.hg.1 | ENST00000567548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010522.hg.1 | ENST00000567548.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029780.hg.1 | TC01001915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029780.hg.1 | ENST00000366646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029780.hg.1 | ENST00000366647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029780.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188221.hg.1 | TC0100011958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000366647.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000416000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000436239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000469332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | ENST00000492459.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011958.hg.1 | NM_014236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010522.hg.1 | ENST00000567548.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010522.hg.1 | ENST00000567548.2 | Ensembl |

GNPAT
Genomic Map