Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000599949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000599949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
ENST00000599949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009700.hg.1 |
TC19002645.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009700.hg.1 |
ENST00000253237.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009700.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201852.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201852.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201852.hg.1 |
TC1900011775.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900201852.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201852.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000598711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000599949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000599949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
ENST00000599949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011775.hg.1 |
NM_031485.3 |
Refseq |