Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000523392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
ENST00000523392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014393.hg.1 |
TC08001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014393.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014393.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000339382.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000523392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
ENST00000523392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_001015508.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010048.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168412.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168412.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168412.hg.1 |
ENST00000475541.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168412.hg.1 |
NM_013357.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168412.hg.1 |
TC0800010048.hg.1 |
Clariom D TC |