Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043551.hg.1 |
TC02002703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043551.hg.1 |
ENST00000374412.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043551.hg.1 |
ENST00000392214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043551.hg.1 |
ENST00000449792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043551.hg.1 |
ENST00000454776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043551.hg.1 |
NM_001039845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043550.hg.1 |
TC02002703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043550.hg.1 |
ENST00000374412.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043550.hg.1 |
ENST00000392214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043550.hg.1 |
ENST00000449792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043550.hg.1 |
ENST00000454776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043550.hg.1 |
NM_001039845.2 |
Refseq |