Assay Details
Gene Symbol: CEBPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005760.2 | NP_005751.2 | 4 - 5 | 2089 | 77 | |
GenBank mRNA | AK312322.1 | - | 4 - 5 | 2088 | 77 | |
BC034475.2 | - | 4 - 5 | 2093 | 4822708 | 77 | |
M37197.1 | - | 4 - 5 | 2070 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001741.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
poly(A) RNA binding
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02028050.hg.1 | TC02001741.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02028050.hg.1 | ENST00000234170.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02028050.hg.1 | ENST00000545744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02028050.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | ENST00000234170.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | ENST00000619061.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | NM_005760.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200189497.hg.1 | TC0200012260.hg.1 | Clariom D TC |
