Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006644.hg.1 |
ENST00000415816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006644.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006644.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006644.hg.1 |
TC17000545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000396722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000396722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000396722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000536969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000536969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000536969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000246911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000396722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000396722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000396722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000534876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000536969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000536969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000536969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000538473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
ENST00000546325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007931.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
ENST00000415816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
ENST00000438323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
NM_005533.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157980.hg.1 |
TC1700007931.hg.1 |
Clariom D TC |