Assay Details
| Gene Symbol: ERI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL137679.1 | - | 7 - 8 | 1323 | 154 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000250263.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000518663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000518663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000518663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000518663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000518663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000519292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520332.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000520684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000521844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000521844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000521844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000521844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000521844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000522612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | ENST00000523898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006663.hg.1 | NM_153332.3 | Refseq |

ERI1
Genomic Map