Assay Details
| Gene Symbol: PINX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017884.5 | NP_060354.4 | 5 - 6 | 530 | 89 | |
| GenBank mRNA | AF418553.1 | - | 5 - 6 | 519 | 89 | |
| AJ344104.1 | - | 5 - 6 | 519 | 89 | ||
| AK313715.1 | - | 5 - 6 | 468 | 89 | ||
| AY523566.1 | - | 5 - 6 | 412 | 89 | ||
| BC015479.1 | - | 5 - 6 | 476 | 3914396 | 89 | |
| BC093762.1 | - | 5 - 6 | 420 | 7939607 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08002611.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
telomere maintenance via telomerase
mitotic metaphase plate congression
negative regulation of cell proliferation
negative regulation of protein ubiquitination
regulation of protein stability
negative regulation of telomere maintenance via telomerase
regulation of telomerase activity
negative regulation of telomerase activity
protein localization to chromosome, telomeric region
protein localization to nucleolus
negative regulation of telomere maintenance via telomere lengthening
positive regulation of telomeric DNA binding
positive regulation of protein localization to nucleolus
protein binding
telomerase inhibitor activity
telomerase RNA binding
telomere maintenance via telomerase
mitotic metaphase plate congression
negative regulation of cell proliferation
negative regulation of protein ubiquitination
regulation of protein stability
negative regulation of telomere maintenance via telomerase
regulation of telomerase activity
negative regulation of telomerase activity
protein localization to chromosome, telomeric region
protein localization to nucleolus
negative regulation of telomere maintenance via telomere lengthening
positive regulation of telomeric DNA binding
positive regulation of protein localization to nucleolus
protein binding
telomerase inhibitor activity
telomerase RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012386.hg.1 | NR_031711.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012224.hg.1 | TC08002611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012224.hg.1 | ENST00000314787.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012224.hg.1 | ENST00000426190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012224.hg.1 | ENST00000519088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012224.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012225.hg.1 | TC08002611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012225.hg.1 | ENST00000314787.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012225.hg.1 | ENST00000426190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012225.hg.1 | ENST00000519088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012225.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186601.hg.1 | TC0800012386.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000314787.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000517607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000519088.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000520018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000523559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | ENST00000524114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_001284356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NM_017884.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012386.hg.1 | NR_031711.1 | Refseq |

PINX1
Genomic Map