Assay Details
Gene Symbol: ZFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015174.1 | NP_055989.1 | 15 - 16 | 2352 | 147 | |
XM_011527831.2 | XP_011526133.1 | 15 - 16 | 2354 | 147 | ||
GenBank mRNA | AB029009.1 | - | 14 - 15 | 2230 | 147 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001052.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591712.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014358.hg.1 | ENST00000262961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19014358.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014358.hg.1 | TC19001052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19014359.hg.1 | ENST00000262961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19014359.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014359.hg.1 | TC19001052.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900174295.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174295.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174295.hg.1 | TC1900009311.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900174295.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174295.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000438164.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000439086.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000586578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000587212.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591712.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000591965.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | ENST00000592807.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_001145640.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009311.hg.1 | NR_027149.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174294.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174294.hg.1 | NM_015174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174294.hg.1 | TC1900009311.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900174294.hg.1 | ENST00000262961.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174294.hg.1 | NM_015174.1 | Refseq |
