Assay Details
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152490.4 | NP_689703.1 | 10 - 11 | 1536 | 69 | |
XM_006711749.3 | XP_006711812.1 | 10 - 11 | 1478 | 69 | ||
XM_017000394.1 | XP_016855883.1 | 11 - 12 | 1659 | 69 | ||
GenBank mRNA | AB209046.1 | - | 9 - 10 | 1090 | 69 | |
AK311626.1 | - | 5 - 6 | 808 | 69 | ||
BC029564.1 | - | 10 - 11 | 1536 | 4826983 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003994.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01060451.hg.1 | TC01003994.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01060451.hg.1 | ENST00000366599.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060451.hg.1 | ENST00000366600.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060451.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01060452.hg.1 | TC01003994.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01060452.hg.1 | ENST00000366599.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060452.hg.1 | ENST00000366600.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060452.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234504.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234504.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234504.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234504.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234504.hg.1 | TC0100017828.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234503.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234503.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234503.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234503.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234503.hg.1 | TC0100017828.hg.1 | Clariom D TC |
