Assay Details
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277155.2 | NP_001264084.1 | 4 - 5 | 710 | 122 | |
NM_152490.4 | NP_689703.1 | 3 - 4 | 587 | 122 | ||
XM_006711749.3 | XP_006711812.1 | 3 - 4 | 529 | 122 | ||
XM_017000394.1 | XP_016855883.1 | 4 - 5 | 710 | 122 | ||
XM_017000395.1 | XP_016855884.1 | 4 - 5 | 663 | 122 | ||
GenBank mRNA | AB209046.1 | - | 2 - 3 | 141 | 122 | |
BC016974.1 | - | 4 - 5 | 651 | 122 | ||
BC029564.1 | - | 3 - 4 | 587 | 4826983 | 122 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003994.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01060468.hg.1 | TC01003994.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01060468.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060468.hg.1 | ENST00000366599.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060468.hg.1 | ENST00000366600.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060468.hg.1 | ENST00000478199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060468.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234520.hg.1 | TC0100017828.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000313984.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000366600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000461994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000462374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000477694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000494378.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | ENST00000612859.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_001277155.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017828.hg.1 | NM_152490.4 | Refseq |
