Assay Details
Gene Symbol: ZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021186.4 | NP_067009.1 | 6 - 7 | 1122 | 105 | |
GenBank mRNA | AK314151.1 | - | 6 - 7 | 1122 | 105 | |
BC069521.1 | - | 6 - 7 | 835 | 7262334 | 105 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01004010.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of humoral immune response
binding of sperm to zona pellucida
protein kinase A signaling
intracellular signal transduction
positive regulation of T cell proliferation
positive regulation of protein kinase activity
acrosomal vesicle exocytosis
protein kinase C signaling
positive regulation of acrosome reaction
negative regulation of binding of sperm to zona pellucida
signal transducer activity
acrosin binding
positive regulation of humoral immune response
binding of sperm to zona pellucida
protein kinase A signaling
intracellular signal transduction
positive regulation of T cell proliferation
positive regulation of protein kinase activity
acrosomal vesicle exocytosis
protein kinase C signaling
positive regulation of acrosome reaction
negative regulation of binding of sperm to zona pellucida
signal transducer activity
acrosin binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01060708.hg.1 | TC01004010.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01060708.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01060708.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017878.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | ENST00000366570.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | ENST00000611898.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | NM_021186.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100234876.hg.1 | TC0100017878.hg.1 | Clariom D TC |
