Assay Details
| Gene Symbol: MROH3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ983818.1 | - | 11 - 12 | 1529 | 80 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |

MROH3P
Genomic Map