Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007572.hg.1 |
ENST00000361722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007572.hg.1 |
ENST00000537587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007572.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007572.hg.1 |
TC17000612.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008134.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159889.hg.1 |
TC1700008134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000361722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000537587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000578982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
ENST00000622396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
NM_014726.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159888.hg.1 |
TC1700008134.hg.1 |
Clariom D TC |