Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015717.hg.1 |
TC02001056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015717.hg.1 |
ENST00000331462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015717.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015722.hg.1 |
TC02001056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015722.hg.1 |
ENST00000331462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015722.hg.1 |
ENST00000375170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015722.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015718.hg.1 |
TC02001056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015718.hg.1 |
ENST00000331462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015718.hg.1 |
ENST00000375170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015718.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
ENST00000610524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010053.hg.1 |
NR_106991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
ENST00000610524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010053.hg.1 |
NR_106991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170554.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170554.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170554.hg.1 |
ENST00000331462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170554.hg.1 |
NM_024501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170554.hg.1 |
TC0200010053.hg.1 |
Clariom D TC |