Assay Details
| Gene Symbol: MPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001932.4 | NP_001923.2 | 13 - 14 | 1144 | 70 | |
| XM_006721915.2 | XP_006721978.1 | 14 - 15 | 1332 | 70 | ||
| XM_011524828.2 | XP_011523130.1 | 13 - 14 | 1227 | 70 | ||
| XM_017024656.1 | XP_016880145.1 | 13 - 14 | 1360 | 70 | ||
| XM_017024657.1 | XP_016880146.1 | 12 - 13 | 1036 | 70 | ||
| GenBank mRNA | AK313045.1 | - | 13 - 14 | 1111 | 70 | |
| AM050144.1 | - | 11 - 12 | 975 | 70 | ||
| BC056865.1 | - | 12 - 13 | 1058 | 5178132 | 70 | |
| U37707.1 | - | 11 - 12 | 1305 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

MPP3
Genomic Map