Assay Details
Gene Symbol: NAGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005261616.4 | XP_005261673.1 | 1 - 2 | 98 | 54 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005261616.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000396398.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000402937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | ENST00000403363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008856.hg.1 | NM_000262.2 | Refseq |
