Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
TC03001998.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000284483.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000341852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000357327.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000369326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000436636.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000460047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000470834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000475336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000488470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
ENST00000538048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161562.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161564.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161565.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_001161566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035730.hg.1 |
NM_015028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000284483.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000341852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000357327.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000436636.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000460047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000470834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000475336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000488470.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161561.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161564.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_015028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000284483.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000341852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000357327.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000436636.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000460047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000470834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000475336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
ENST00000488470.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161561.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161562.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161564.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161565.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_001161566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
NM_015028.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198069.hg.1 |
TC0300013108.hg.1 |
Clariom D TC |