Assay Details
| Gene Symbol: TNIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161561.2 | NP_001155033.1 | 19 - 20 | 2636 | 83 | |
| NM_001161563.2 | NP_001155035.1 | 19 - 20 | 2558 | 83 | ||
| NM_001161565.2 | NP_001155037.1 | 18 - 19 | 2471 | 83 | ||
| NM_015028.3 | NP_055843.1 | 20 - 21 | 2723 | 83 | ||
| GenBank mRNA | AB011123.2 | - | 20 - 21 | 2644 | 83 | |
| AF172264.1 | - | 20 - 21 | 2378 | 83 | ||
| AF172265.1 | - | 19 - 20 | 2291 | 83 | ||
| AF172266.1 | - | 19 - 20 | 2213 | 83 | ||
| AF172268.1 | - | 18 - 19 | 2126 | 83 | ||
| AK291056.1 | - | 18 - 19 | 2471 | 83 | ||
| BC150256.1 | - | 20 - 21 | 2644 | 9007184 | 83 | |
More Information
Set Membership: |
|

TNIK
Genomic Map