| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000314607.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000364672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000441447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000441447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000441447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000441447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000457946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000470175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000470175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000470175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000470175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000482131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000482131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000482131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000482131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000492456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000492456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000492456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
ENST00000492456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007765.hg.1 |
NM_005095.2 |
Refseq |