Assay Details
| Gene Symbol: ZMYM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005095.2 | NP_005086.2 | 1 - 2 | 119 | 104 | |
| GenBank mRNA | AK308320.1 | - | 1 - 2 | 119 | 104 | |
| BG499626.1 | - | 1 - 2 | 120 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000456.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_005095.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding
regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000314607.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000364672.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000441447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000441447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000441447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000441447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000457946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000470175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000470175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000470175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000470175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000482131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000482131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000482131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000482131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000492456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000492456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000492456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | ENST00000492456.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007765.hg.1 | NM_005095.2 | Refseq |

ZMYM4
Genomic Map