Assay Details
| Gene Symbol: ZMYM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005095.2 | NP_005086.2 | 21 - 22 | 3331 | 105 | |
| XM_005271328.4 | XP_005271385.1 | 21 - 22 | 3610 | 105 | ||
| XM_005271331.2 | XP_005271388.1 | 18 - 19 | 2576 | 105 | ||
| XM_011542421.2 | XP_011540723.1 | 21 - 22 | 3257 | 105 | ||
| XM_011542423.2 | XP_011540725.1 | 22 - 23 | 3662 | 105 | ||
| XM_011542425.2 | XP_011540727.1 | 19 - 20 | 2616 | 105 | ||
| XM_017002803.1 | XP_016858292.1 | 22 - 23 | 3384 | 105 | ||
| XM_017002804.1 | XP_016858293.1 | 22 - 23 | 3324 | 105 | ||
| XM_017002805.1 | XP_016858294.1 | 17 - 18 | 2822 | 105 | ||
| GenBank mRNA | AB007885.1 | - | 17 - 18 | 2451 | 105 | |
| AK309705.1 | - | 20 - 21 | 2991 | 105 | ||
| BC012093.1 | - | 19 - 20 | 2910 | 105 | ||
| BC127113.1 | - | 22 - 23 | 3337 | 40125110 | 105 | |
| BC127114.1 | - | 18 - 19 | 2520 | 40125115 | 105 | |
| BX647376.1 | - | 17 - 18 | 2562 | 105 | ||
| BX648067.1 | - | 12 - 13 | 1625 | 105 | ||
| CR936741.1 | - | 20 - 21 | 3195 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000456.hg.1
Set Membership: |
|

ZMYM4
Genomic Map