Assay Details
| Gene Symbol: MAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080600.2 | NP_542167.1 | 11 - 12 | 1962 | 71 | |
| GenBank mRNA | BC093045.1 | - | 11 - 12 | 1914 | 30396005 | 71 |
| X98405.1 | - | 3 - 4 | 184 | 71 | ||
More Information
Set Membership: |
|

MAG
Genomic Map