Assay Details
| Gene Symbol: TMEM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530112.2 | XP_011528414.2 | 8 - 9 | 1240 | 70 | |
| XM_017028756.1 | XP_016884245.1 | 8 - 9 | 1170 | 70 | ||
| XM_017028757.1 | XP_016884246.1 | 7 - 8 | 958 | 70 | ||
More Information
Set Membership: |
|

TMEM184B
Genomic Map