Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC004500.2 | - | 1 - 2 | 47 | 3833318 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC004500.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153691.hg.1 | TC0900007977.hg.1 | Clariom D TC |
