Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032310.4 | NP_115686.3 | 4 - 5 | 588 | 81 | |
NR_134466.1 | 4 - 5 | 599 | 81 | |||
NR_134467.1 | 5 - 6 | 690 | 81 | |||
GenBank mRNA | BC004500.2 | - | 5 - 6 | 457 | 3833318 | 81 |
BC091476.1 | - | 4 - 5 | 442 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000442.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004727.hg.1 | TC09000442.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09004727.hg.1 | ENST00000375464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004727.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004727.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153710.hg.1 | TC0900007977.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000375464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000466929.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000468781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000475574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000488630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000490488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000495641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | ENST00000498243.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007977.hg.1 | NM_032310.4 | Refseq |
