Assay Details
| Gene Symbol: PRPF38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032864.3 | NP_116253.2 | 3 - 4 | 610 | 164 | |
| XM_011542315.2 | XP_011540617.1 | 3 - 4 | 610 | 164 | ||
| GenBank mRNA | AK027842.1 | - | 2 - 3 | 184 | 164 | |
| AK092038.1 | - | 3 - 4 | 593 | 164 | ||
| BC105004.1 | - | 3 - 4 | 451 | 8144007 | 164 | |
| BC105006.1 | - | 3 - 4 | 451 | 8144009 | 164 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000641.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010487.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010487.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010487.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010487.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010486.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010486.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010486.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159255.hg.1 | TC0100008315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159254.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159254.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159254.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159254.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159254.hg.1 | TC0100008315.hg.1 | Clariom D TC |

PRPF38A
Genomic Map