Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021037.hg.1 |
TC09001654.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021037.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021037.hg.1 |
ENST00000427355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021037.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021035.hg.1 |
TC09001654.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021035.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021035.hg.1 |
ENST00000427355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021035.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011709.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180102.hg.1 |
TC0900011709.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
ENST00000351698.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
ENST00000473604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
ENST00000474192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
NM_000113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180103.hg.1 |
TC0900011709.hg.1 |
Clariom D TC |