Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006611.hg.1 |
TC08000476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006611.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006611.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006610.hg.1 |
TC08000476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006610.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08006610.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154770.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800154770.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154770.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800154770.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154770.hg.1 |
TC0800007945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800154772.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800154772.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154772.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800154772.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154772.hg.1 |
TC0800007945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800154771.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800154771.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154771.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800154771.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800154771.hg.1 |
TC0800007945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
ENST00000523207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007945.hg.1 |
NM_004770.2 |
Refseq |