Assay Details
| Gene Symbol: CENPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318521.1 | NP_001305450.1 | 16 - 17 | 1746 | 128 | |
| NM_006733.3 | NP_006724.2 | 15 - 16 | 1840 | 128 | ||
| XM_005262111.2 | XP_005262168.1 | 16 - 17 | 1710 | 128 | ||
| XM_011530894.2 | XP_011529196.1 | 16 - 17 | 1739 | 128 | ||
| XM_011530895.2 | XP_011529197.1 | 17 - 18 | 2026 | 128 | ||
| XM_011530897.2 | XP_011529199.1 | 17 - 18 | 1901 | 128 | ||
| XM_011530899.2 | XP_011529201.1 | 15 - 16 | 1707 | 128 | ||
| XM_017029383.1 | XP_016884872.1 | 16 - 17 | 1739 | 128 | ||
| GenBank mRNA | AK302986.1 | - | 16 - 17 | 1739 | 128 | |
| M78295.1 | - | 3 - 4 | 184 | 128 | ||
| X97249.1 | - | 15 - 16 | 1839 | 128 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006145.hg.1 | TC0X000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006145.hg.1 | ENST00000218507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006145.hg.1 | ENST00000372927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006145.hg.1 | ENST00000423383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006145.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | ENST00000372927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | ENST00000423383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | NM_006733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153898.hg.1 | TC0X00007936.hg.1 | Clariom D TC |

CENPI
Genomic Map