Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092414.1 | - | 3 - 4 | 2383 | 74 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000343242.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000403150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | ENST00000406174.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006520.hg.1 | NM_002360.3 | Refseq |

MAFK
Genomic Map