Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001351.hg.1 |
TC03000062.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001351.hg.1 |
ENST00000254488.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001351.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145771.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145771.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145771.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145771.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145771.hg.1 |
TC0300006596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000254488.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000454147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
ENST00000464828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006596.hg.1 |
NM_014229.2 |
Refseq |