Assay Details
Gene Symbol: TBKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005257861.4 | XP_005257918.1 | 1 - 2 | 420 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005257861.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000361722.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000537587.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000578982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | ENST00000622396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008134.hg.1 | NM_014726.2 | Refseq |
