Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024308.3 | NP_077284.2 | 3 - 4 | 668 | 106 | |
| XM_005257658.2 | XP_005257715.1 | 3 - 4 | 673 | 106 | ||
| XM_011525233.2 | XP_011523535.1 | 2 - 3 | 408 | 106 | ||
| GenBank mRNA | AK026196.1 | - | 3 - 4 | 459 | 106 | |
| AK315735.1 | - | 3 - 4 | 589 | 106 | ||
| AY358712.1 | - | 3 - 4 | 565 | 106 | ||
| BC002731.2 | - | 3 - 4 | 554 | 3631177 | 106 | |
| CR457356.1 | - | 2 - 3 | 217 | 106 | ||
| LC110386.1 | - | 3 - 4 | 454 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000439.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17005227.hg.1 | ENST00000251312.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005227.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005227.hg.1 | TC17000439.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700155120.hg.1 | TC1700007641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000610443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000611337.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000612538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000615432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000617959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000618403.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | ENST00000621871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007641.hg.1 | NM_024308.3 | Refseq |

DHRS11
Genomic Map