Assay Details
| Gene Symbol: GNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021629.3 | NP_067642.1 | 6 - 7 | 713 | 120 | |
| XM_005247692.2 | XP_005247749.1 | 6 - 7 | 634 | 120 | ||
| XM_006713721.2 | XP_006713784.1 | 6 - 7 | 644 | 120 | ||
| GenBank mRNA | AF300648.1 | - | 5 - 6 | 433 | 120 | |
| AK001890.1 | - | 6 - 7 | 713 | 120 | ||
| AK290571.1 | - | 6 - 7 | 644 | 120 | ||
| AK297174.1 | - | 6 - 7 | 510 | 120 | ||
| AK310009.1 | - | 2 - 3 | 200 | 120 | ||
| BC000873.1 | - | 6 - 7 | 523 | 3460570 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002038.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297174.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03036086.hg.1 | TC03002038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036086.hg.1 | ENST00000232564.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036086.hg.1 | ENST00000468623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036086.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000595265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198836.hg.1 | TC0300013264.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000595265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |

GNB4
Genomic Map