Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
TC03000642.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
ENST00000291478.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
ENST00000360013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
ENST00000393496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
ENST00000428018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
ENST00000459915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
NM_001024660.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011657.hg.1 |
NM_007064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000291478.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000354186.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000360013.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000393496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000459915.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
NM_001024660.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
NM_007064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000291478.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000354186.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000360013.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000393496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
ENST00000459915.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
NM_001024660.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
NM_007064.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161259.hg.1 |
TC0300008594.hg.1 |
Clariom D TC |