Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000355681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000420781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000443566.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000446176.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000449089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000462766.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000478129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
NM_015033.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000355681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000420781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000443566.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000446176.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000449089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000462766.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
ENST00000478129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
NM_015033.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180164.hg.1 |
TC0900011713.hg.1 |
Clariom D TC |