Assay Details
| Gene Symbol: PRPF38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032864.3 | NP_116253.2 | 1 - 2 | 330 | 92 | |
| XM_011542315.2 | XP_011540617.1 | 1 - 2 | 330 | 92 | ||
| GenBank mRNA | AK092038.1 | - | 1 - 2 | 313 | 92 | |
| BC105004.1 | - | 1 - 2 | 171 | 8144007 | 92 | |
| BC105006.1 | - | 1 - 2 | 171 | 8144009 | 92 | |
| DA055001.1 | - | 1 - 2 | 330 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000641.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010481.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010481.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010481.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010481.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006712.hg.1 | ENST00000451976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010480.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010480.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010480.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010480.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010483.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010483.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010483.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006900.hg.1 | ENST00000438179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010482.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010482.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010482.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010482.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010478.hg.1 | TC01000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010478.hg.1 | ENST00000257181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010478.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159248.hg.1 | TC0100008315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159250.hg.1 | TC0100008315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000006712.hg.1 | ENST00000451976.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159245.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159245.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159245.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159245.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159245.hg.1 | TC0100008315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000006900.hg.1 | ENST00000438179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159249.hg.1 | TC0100008315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159247.hg.1 | TC0100008315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000474048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008315.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | ENST00000257181.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | ENST00000487160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | NM_032864.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159251.hg.1 | TC0100008315.hg.1 | Clariom D TC |

PRPF38A
Genomic Map